Assay Details
| Gene Symbol: Atp1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178405.3 | NP_848492.1 | 1 - 2 | 186 | 55 | |
| GenBank mRNA | AK052863.1 | - | 1 - 2 | 122 | 55 | |
| AK129212.1 | - | 1 - 2 | 99 | 55 | ||
| AK137602.1 | - | 1 - 2 | 122 | 55 | ||
| AK147331.1 | - | 1 - 2 | 118 | 55 | ||
| AK148775.1 | - | 1 - 2 | 118 | 55 | ||
| AK161840.1 | - | 1 - 2 | 118 | 55 | ||
| AK164509.1 | - | 1 - 2 | 122 | 55 | ||
| AK168703.1 | - | 1 - 2 | 118 | 55 | ||
| BC036127.1 | - | 1 - 2 | 110 | 4955003 | 55 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK137602.1, AK147331.1, AK161840.1, AK164509.1, AK168703.1, BC036127.1, NM_178405.3 transcripts.
Set Membership: |
|

Atp1a2
Genomic Map