Assay Details
| Gene Symbol: Atp1a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013734.1 | NP_038762.1 | 11 - 12 | 1590 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transport
ion transport
potassium ion transport
sodium ion transport
spermatogenesis
fertilization
sperm motility
regulation of cellular pH
sodium ion transmembrane transport
regulation of membrane potential
potassium ion transmembrane transport
nucleotide binding
sodium:potassium-exchanging ATPase activity
ATP binding
hydrolase activity
metal ion binding
transport
ion transport
potassium ion transport
sodium ion transport
spermatogenesis
fertilization
sperm motility
regulation of cellular pH
sodium ion transmembrane transport
regulation of membrane potential
potassium ion transmembrane transport
nucleotide binding
sodium:potassium-exchanging ATPase activity
ATP binding
hydrolase activity
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2184123 | NM_013734.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2009787 | NM_013734.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 454739 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 489125 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 273765 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 407425 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 780511 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 378355 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 234100 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 969867 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 141800 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 592100 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 429132 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 865355 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 259824 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 835432 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 682402 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 531034 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 520799 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 237374 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 669689 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 834205 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 180058 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 768271 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 123278 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 44891 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 19159 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2184123 | NM_013734.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009787 | NM_013734.1 | mouse_transcriptome |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10360264 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360249 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360255 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360269 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360266 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360252 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360265 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360268 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360256 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360267 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360254 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360259 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360251 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360260 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360262 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360257 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360261 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360258 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360253 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360263 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360250 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437321_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10360248 | - | - |
| Mouse Genome 430 2.0 Array | 1437321_at | - | - |
| Mouse Genome 430 2.0 Array | 1437321_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100029280.mm.1 | NM_013734.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029280.mm.1 | NM_013734.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029280.mm.1 | TC0100003565.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1437321_PM_at | - | - |
| Mouse Expression Set 430 | 1437321_at | - | - |
| Murine Genome U74v2 Set | 166379_PM_at | - | - |
| Murine Genome U74v2 Set | 166379_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437321_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437321_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 669398 | - | - |

Atp1a4
Genomic Map