Assay Details
| Gene Symbol: Atp1a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013734.1 | NP_038762.1 | 19 - 20 | 2840 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp1a4| Gene Symbol: Atp1a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013734.1 | NP_038762.1 | 19 - 20 | 2840 | 125 | |
Set Membership: |
|