Assay Details
| Gene Symbol: Iqca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029122.2 | NP_083398.2 | 9 - 10 | 1441 | 115 | |
| GenBank mRNA | AK139726.1 | - | 2 - 3 | 296 | 115 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK139726.1 transcripts.
Set Membership: |
|

Iqca
Genomic Map