Assay Details
| Gene Symbol: Iqca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029122.2 | NP_083398.2 | 15 - 16 | 2096 | 62 | |
| GenBank mRNA | AK139726.1 | - | 8 - 9 | 951 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Iqca
Genomic Map