Assay Details
| Gene Symbol: Iqca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029122.2 | NP_083398.2 | 3 - 4 | 678 | 71 | |
| GenBank mRNA | AK015512.1 | - | 3 - 4 | 679 | 71 | |
| AK016353.1 | - | 3 - 4 | 679 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Iqca
Genomic Map