Assay Details
| Gene Symbol: Iqca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029122.2 | NP_083398.2 | 6 - 7 | 1148 | 80 | |
| GenBank mRNA | AK015512.1 | - | 6 - 7 | 1149 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Iqca
Genomic Map