Assay Details
| Gene Symbol: Dis3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172157.1 | NP_001165628.1 | 10 - 11 | 1322 | 73 | |
| NM_153530.2 | NP_705758.1 | 9 - 10 | 1280 | 73 | ||
| GenBank mRNA | AK029652.1 | - | 10 - 11 | 1336 | 73 | |
| AK031180.1 | - | 10 - 11 | 1305 | 73 | ||
| AK213947.1 | - | 2 - 3 | 216 | 73 | ||
| AK214096.1 | - | 2 - 3 | 213 | 73 | ||
| BC036177.1 | - | 9 - 10 | 1241 | 5004341 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dis3l2
Genomic Map