Assay Details
| Gene Symbol: Dis3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172157.1 | NP_001165628.1 | 16 - 17 | 2123 | 84 | |
| NM_153530.2 | NP_705758.1 | 15 - 16 | 2081 | 84 | ||
| GenBank mRNA | AK029652.1 | - | 16 - 17 | 2137 | 84 | |
| AK031180.1 | - | 16 - 17 | 2106 | 84 | ||
| BC036177.1 | - | 15 - 16 | 2042 | 5004341 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dis3l2
Genomic Map