Assay Details
| Gene Symbol: Dis3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK029652.1 | - | 6 - 7 | 589 | 94 | |
| AK040613.1 | - | 7 - 8 | 631 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK029652.1 transcripts.
Set Membership: |
|

Dis3l2
Genomic Map