Assay Details
| Gene Symbol: Slc26a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021353.3 | NP_067328.1 | 6 - 7 | 927 | 73 | |
| GenBank mRNA | AF136751.1 | - | 6 - 7 | 845 | 73 | |
| AK018566.1 | - | 6 - 7 | 878 | 73 | ||
| AK035716.1 | - | 4 - 5 | 880 | 73 | ||
| BC022639.1 | - | 3 - 4 | 447 | 73 | ||
| BC037066.1 | - | 6 - 7 | 923 | 73 | ||
| BC139271.1 | - | 5 - 6 | 762 | 8862293 | 73 | |
| BC139273.1 | - | 5 - 6 | 762 | 8862295 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc26a3
Genomic Map