Assay Details
| Gene Symbol: Slc26a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021353.3 | NP_067328.1 | 7 - 8 | 1084 | 79 | |
| GenBank mRNA | AF136751.1 | - | 7 - 8 | 1002 | 79 | |
| BC139271.1 | - | 6 - 7 | 919 | 8862293 | 79 | |
| BC139273.1 | - | 6 - 7 | 919 | 8862295 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc26a3
Genomic Map