Assay Details
| Gene Symbol: Havcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166631.1 | NP_001160103.1 | 4 - 5 | 719 | 73 | |
| NM_001166632.1 | NP_001160104.1 | 4 - 5 | 603 | 73 | ||
| GenBank mRNA | AF399830.1 | - | 3 - 4 | 549 | 73 | |
| AK133466.1 | - | 4 - 5 | 721 | 73 | ||
| BC053400.1 | - | 4 - 5 | 611 | 30033601 | 73 | |
More Information
Set Membership: |
|

Havcr1
Genomic Map