Assay Details
| Gene Symbol: Gnb2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008143.3 | NP_032169.1 | 3 - 4 | 535 | 68 | |
| GenBank mRNA | AK002493.1 | - | 3 - 4 | 524 | 68 | |
| AK012242.1 | - | 2 - 3 | 322 | 68 | ||
| AK017772.1 | - | 3 - 4 | 536 | 68 | ||
| AK051226.1 | - | 3 - 4 | 536 | 68 | ||
| AK159797.1 | - | 3 - 4 | 536 | 68 | ||
| AK160528.1 | - | 3 - 4 | 506 | 68 | ||
| AK160739.1 | - | 3 - 4 | 533 | 68 | ||
| AK161234.1 | - | 3 - 4 | 536 | 68 | ||
| AK166417.1 | - | 3 - 4 | 524 | 68 | ||
| AK166786.1 | - | 3 - 4 | 538 | 68 | ||
| AK166800.1 | - | 3 - 4 | 539 | 68 | ||
| AK166889.1 | - | 2 - 3 | 176 | 68 | ||
| AK166945.1 | - | 3 - 4 | 536 | 68 | ||
| AK168086.1 | - | 3 - 4 | 536 | 68 | ||
| AK168096.1 | - | 3 - 4 | 536 | 68 | ||
| AK168196.1 | - | 3 - 4 | 536 | 68 | ||
| AK168349.1 | - | 3 - 4 | 567 | 68 | ||
| AK169612.1 | - | 3 - 4 | 536 | 68 | ||
| BC020322.1 | - | 3 - 4 | 512 | 68 | ||
| BC036295.1 | - | 1 - 2 | 75 | 68 | ||
| BC046760.1 | - | 3 - 4 | 520 | 5693613 | 68 | |
| D29802.1 | - | 3 - 4 | 529 | 68 | ||
| X75313.1 | - | 3 - 4 | 535 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gnb2l1
Genomic Map