Assay Details
Gene Symbol: Mgarp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026358.3 | NP_080634.2 | 1 - 2 | 124 | 115 | |
GenBank mRNA | AF228502.1 | - | 1 - 2 | 103 | 115 | |
AK006339.1 | - | 1 - 2 | 86 | 115 | ||
AK016342.1 | - | 1 - 2 | 97 | 114 | ||
AK151401.1 | - | 1 - 2 | 118 | 115 | ||
BC012511.1 | - | 1 - 2 | 117 | 4500810 | 115 | |
BC049556.1 | - | 1 - 2 | 86 | 6772062 | 115 | |
BY328038.1 | - | 1 - 2 | 124 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
