Assay Details
| Gene Symbol: Pogk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142948.1 | NP_001136420.1 | 1 - 2 | 116 | 89 | |
| GenBank mRNA | AK048752.1 | - | 1 - 2 | 75 | 89 | |
| BC072587.1 | - | 70 | 6829796 | 89 | ||
More Information
Set Membership: |
|

Pogk
Genomic Map