Assay Details
| Gene Symbol: Edil3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037987.3 | NP_001033076.1 | 4 - 5 | 858 | 99 | |
| NM_010103.4 | NP_034233.1 | 3 - 4 | 828 | 99 | ||
| GenBank mRNA | AF031524.1 | - | 4 - 5 | 966 | 99 | |
| AF031525.1 | - | 4 - 5 | 897 | 99 | ||
| AK036117.1 | - | 4 - 5 | 819 | 99 | ||
| AK046885.1 | - | 4 - 5 | 859 | 99 | ||
| AK081003.1 | - | 3 - 4 | 660 | 99 | ||
| AK084215.1 | - | 4 - 5 | 713 | 99 | ||
| AK135220.1 | - | 4 - 5 | 822 | 99 | ||
| AK139093.1 | - | 4 - 5 | 820 | 99 | ||
| AK141907.1 | - | 4 - 5 | 842 | 99 | ||
| AK157810.1 | - | 4 - 5 | 843 | 99 | ||
| BC056386.1 | - | 4 - 5 | 758 | 6854497 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Edil3
Genomic Map