Assay Details
| Gene Symbol: Edil3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037987.3 | NP_001033076.1 | 7 - 8 | 1318 | 153 | |
| NM_010103.4 | NP_034233.1 | 6 - 7 | 1288 | 153 | ||
| GenBank mRNA | AF031524.1 | - | 7 - 8 | 1426 | 153 | |
| AK036117.1 | - | 7 - 8 | 1279 | 153 | ||
| AK046885.1 | - | 7 - 8 | 1319 | 153 | ||
| AK081003.1 | - | 6 - 7 | 1120 | 153 | ||
| AK139093.1 | - | 7 - 8 | 1280 | 153 | ||
| AK141907.1 | - | 7 - 8 | 1302 | 153 | ||
| BC056386.1 | - | 7 - 8 | 1218 | 6854497 | 153 | |
More Information
Updated Mapping Information
Set Membership: |
|

Edil3
Genomic Map