Assay Details
| Gene Symbol: Edil3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037987.3 | NP_001033076.1 | 2 - 3 | 706 | 63 | |
| GenBank mRNA | AF031524.1 | - | 2 - 3 | 814 | 63 | |
| AF031525.1 | - | 2 - 3 | 745 | 63 | ||
| AK036117.1 | - | 2 - 3 | 667 | 63 | ||
| AK046885.1 | - | 2 - 3 | 707 | 63 | ||
| AK084215.1 | - | 2 - 3 | 561 | 63 | ||
| AK135220.1 | - | 2 - 3 | 670 | 63 | ||
| AK139093.1 | - | 2 - 3 | 668 | 63 | ||
| AK141907.1 | - | 2 - 3 | 690 | 63 | ||
| AK157810.1 | - | 2 - 3 | 691 | 63 | ||
| BC056386.1 | - | 2 - 3 | 606 | 6854497 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Edil3
Genomic Map