Assay Details
| Gene Symbol: Golt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026680.4 | NP_080956.1 | 1 - 2 | 91 | 71 | |
| GenBank mRNA | AK002581.1 | - | 1 - 2 | 99 | 71 | |
| AK141378.1 | - | 1 - 2 | 92 | 71 | ||
| AY707855.1 | - | 1 - 2 | 114 | 71 | ||
| BC024448.1 | - | 1 - 2 | 114 | 4974299 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002581.1, AK141378.1, AY707855.1, BC024448.1, NM_026680.4 transcripts.
Set Membership: |
|

Golt1a
Genomic Map