Assay Details
| Gene Symbol: Ccdc169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290138.1 | NP_001277067.1 | 3 - 4 | 947 | 110 | |
| NM_001290141.1 | NP_001277070.1 | 3 - 4 | 422 | 110 | ||
| NM_177203.3 | NP_796177.2 | 3 - 4 | 326 | 110 | ||
| NR_110776.1 | 3 - 4 | 326 | 110 | |||
| NR_110780.1 | 2 - 3 | 191 | 110 | |||
| NR_110781.1 | 3 - 4 | 225 | 110 | |||
| GenBank mRNA | AK042903.1 | - | 3 - 4 | 949 | 110 | |
| AK076919.1 | - | 3 - 4 | 326 | 110 | ||
| AK160422.1 | - | 3 - 4 | 322 | 110 | ||
| BC089522.1 | - | 3 - 4 | 405 | 110 | ||
| BC150827.1 | - | 2 - 3 | 144 | 110 | ||
| BC171976.1 | - | 2 - 3 | 144 | 110 | ||
| BC171978.1 | - | 3 - 4 | 178 | 110 | ||
| BC171981.1 | - | 2 - 3 | 144 | 110 | ||
| BU962217.1 | - | 328 | 110 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK042903.1, NM_001290138.1, NM_001290141.1 transcripts.
Set Membership: |
|

Ccdc169
Genomic Map