Assay Details
| Gene Symbol: Lgr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033409.3 | NP_001028581.1 | 10 - 11 | 1135 | 71 | |
| GenBank mRNA | AK052873.1 | - | 5 - 6 | 335 | 71 | |
| AK085901.1 | - | 10 - 11 | 1093 | 71 | ||
| AK137380.1 | - | 10 - 11 | 1136 | 71 | ||
| BC141210.1 | - | 10 - 11 | 1087 | 9055822 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lgr6
Genomic Map