Assay Details
| Gene Symbol: Ptpn7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177081.3 | NP_796055.1 | 6 - 7 | 718 | 74 | |
| GenBank mRNA | AK041415.1 | - | 3 - 4 | 2759 | 74 | |
| AK049175.1 | - | 5 - 6 | 569 | 74 | ||
| AK083305.1 | - | 6 - 7 | 718 | 74 | ||
| BC098475.1 | - | 6 - 7 | 772 | 30044369 | 74 | |
| BC138194.1 | - | 5 - 6 | 654 | 8861213 | 74 | |
| BC145190.1 | - | 5 - 6 | 654 | 9053741 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn7
Genomic Map