Assay Details
| Gene Symbol: Dnajc19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026211.2 | NP_001021382.1 | 5 - 6 | 446 | 73 | |
| GenBank mRNA | AK008267.1 | - | 5 - 6 | 390 | 73 | |
| AK076602.1 | - | 405 | 73 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Dnajc19
Genomic Map