Assay Details
| Gene Symbol: Exosc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163570.1 | NP_001157042.1 | 3 - 4 | 451 | 76 | |
| NM_027148.3 | NP_081424.3 | 3 - 4 | 451 | 76 | ||
| GenBank mRNA | AA003291.1 | - | 3 - 4 | 126 | 76 | |
| AJ505005.1 | - | 3 - 4 | 111 | 76 | ||
| AK009584.1 | - | 3 - 4 | 123 | 76 | ||
| AK047257.1 | - | 3 - 4 | 454 | 76 | ||
| AK169719.1 | - | 1 - 2 | 54 | 76 | ||
| AK220227.1 | - | 3 - 4 | 451 | 76 | ||
| BC004769.1 | - | 3 - 4 | 124 | 76 | ||
| BC059089.1 | - | 3 - 4 | 480 | 6509820 | 76 | |
| BC067250.1 | - | 2 - 3 | 94 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Exosc8
Genomic Map