Assay Details
| Gene Symbol: Exosc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163570.1 | NP_001157042.1 | 5 - 6 | 575 | 73 | |
| GenBank mRNA | AA003291.1 | - | 5 - 6 | 250 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exosc8
Genomic Map