Assay Details
| Gene Symbol: Mbnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253709.1 | NP_001240638.1 | 7 - 8 | 983 | 66 | |
| NM_001253713.1 | NP_001240642.1 | 8 - 9 | 1091 | 66 | ||
| NM_020007.3 | NP_064391.2 | 8 - 9 | 2137 | 66 | ||
| GenBank mRNA | AB093240.1 | - | 8 - 9 | 2141 | 66 | |
| AK149405.1 | - | 2 - 3 | 284 | 66 | ||
| AK153827.1 | - | 7 - 8 | 981 | 66 | ||
| AK153848.1 | - | 7 - 8 | 971 | 66 | ||
More Information
Set Membership: |
|

Mbnl1
Genomic Map