Assay Details
| Gene Symbol: Car13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024495.5 | NP_078771.1 | 2 - 3 | 322 | 84 | |
| GenBank mRNA | AF231123.1 | - | 2 - 3 | 272 | 84 | |
| AK010166.1 | - | 2 - 3 | 276 | 84 | ||
| AK150871.1 | - | 2 - 3 | 274 | 84 | ||
| AK150897.1 | - | 2 - 3 | 274 | 84 | ||
| AK151519.1 | - | 2 - 3 | 260 | 84 | ||
| AK151978.1 | - | 2 - 3 | 274 | 84 | ||
| AK152124.1 | - | 2 - 3 | 272 | 84 | ||
| AK153080.1 | - | 2 - 3 | 301 | 84 | ||
| AK153258.1 | - | 2 - 3 | 262 | 84 | ||
| AK153353.1 | - | 2 - 3 | 301 | 84 | ||
| AK162621.1 | - | 2 - 3 | 322 | 84 | ||
| BC064050.1 | - | 2 - 3 | 273 | 949505 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Car13
Genomic Map