Assay Details
| Gene Symbol: Car13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024495.5 | NP_078771.1 | 5 - 6 | 601 | 92 | |
| GenBank mRNA | AF231123.1 | - | 5 - 6 | 551 | 92 | |
| AK010166.1 | - | 5 - 6 | 555 | 92 | ||
| AK150871.1 | - | 5 - 6 | 553 | 92 | ||
| AK150897.1 | - | 5 - 6 | 553 | 92 | ||
| AK151519.1 | - | 5 - 6 | 539 | 92 | ||
| AK151978.1 | - | 5 - 6 | 553 | 92 | ||
| AK152124.1 | - | 5 - 6 | 550 | 92 | ||
| AK153080.1 | - | 5 - 6 | 580 | 92 | ||
| AK153258.1 | - | 5 - 6 | 541 | 92 | ||
| AK153353.1 | - | 5 - 6 | 580 | 92 | ||
| AK162621.1 | - | 5 - 6 | 601 | 92 | ||
| BC064050.1 | - | 5 - 6 | 552 | 949505 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Car13
Genomic Map