Assay Details
| Gene Symbol: 4930440C22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040473.1 | 3 - 4 | 1404 | 130 | ||
| GenBank mRNA | AK015347.1 | - | 3 - 4 | 1407 | 130 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | programmed cell death 1 | Pdcd1 |
Ly101, PD-1, Pdc1 |
-748.4kb | Anti-sense | Chr. 1 - 94038305 - 94052553 on NCBI Build GRCm38 |
| Downstream |
family with sequence similarity 174, member A |
Fam174a |
2310044D20Rik, Fam174, Naa6, Tmem157 |
511.2kb | Sense | Chr. 1 - 95313628 - 95335284 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2419519 | AK015347.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 693517 | - | - |
| Whole Mouse Genome G4122A | A_52_P1027486 | AK015347.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115825 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460666 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460691 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460663 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00049375 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460664 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460616 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460643 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00797931 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460678 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460636 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460635 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460708 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460642 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01091580 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00825768 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01020923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460623 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460649 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460673 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460668 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460646 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01091581 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460707 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460681 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460687 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460694 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00571046 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460639 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460652 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460684 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460655 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460624 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01020925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00825763 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00825765 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460641 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460621 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460629 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01140954 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460647 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01110794 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460682 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00571052 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460699 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460645 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460701 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460698 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00172792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20460674 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1454083_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1454083_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1454083_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100008129.mm.1 | NR_040473.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100008129.mm.1 | NR_040473.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100008129.mm.1 | TC0100000956.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100008126.mm.1 | NR_040473.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100008126.mm.1 | NR_040473.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100008126.mm.1 | TC0100000956.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100008127.mm.1 | NR_040473.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100008127.mm.1 | NR_040473.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100008127.mm.1 | TC0100000956.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100008128.mm.1 | NR_040473.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100008128.mm.1 | NR_040473.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100008128.mm.1 | TC0100000956.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100008131.mm.1 | NR_040473.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100008131.mm.1 | NR_040473.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100008131.mm.1 | TC0100000956.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100008130.mm.1 | NR_040473.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100008130.mm.1 | NR_040473.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100008130.mm.1 | TC0100000956.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1454083_at | - | - |
| Mouse Expression Set 430 | 1454083_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454083_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454083_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 825361 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 104150112 | - | - |

4930440C22Rik
Genomic Map