Assay Details
| Gene Symbol: Lyrm7os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040284.1 | 1 - 2 | 142 | 85 | ||
| GenBank mRNA | AK087691.1 | - | 1 - 2 | 143 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CDC42 small effector 2 | Cdc42se2 |
2810404F18Rik, AA408783, AA536669, SPEC2 |
-32.2kb | Anti-sense | Chr. 11 - 54717415 - 54787703 on NCBI Build GRCm38 |
| Downstream | LYR motif containing 7 | Lyrm7 |
1700024C24Rik, 9330147L21Rik |
5.4kb | Anti-sense | Chr. 11 - 54826866 - 54860907 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Lyrm7os
Genomic Map