Assay Details
| Gene Symbol: Tmem163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028135.2 | NP_082411.1 | 2 - 3 | 710 | 79 | |
| GenBank mRNA | AK011522.1 | - | 2 - 3 | 455 | 79 | |
| AK042650.1 | - | 2 - 3 | 459 | 79 | ||
| AK047006.1 | - | 2 - 3 | 453 | 79 | ||
| BC046234.1 | - | 710 | 6816957 | 79 | ||
| BC059048.1 | - | 710 | 6816957 | 79 | ||
| CA319657.1 | - | 2 - 3 | 710 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem163
Genomic Map