Assay Details
| Gene Symbol: Tmem163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028135.2 | NP_082411.1 | 4 - 5 | 844 | 99 | |
| GenBank mRNA | AK011522.1 | - | 4 - 5 | 589 | 99 | |
| AK042650.1 | - | 4 - 5 | 593 | 99 | ||
| AK047006.1 | - | 4 - 5 | 587 | 99 | ||
| BC046234.1 | - | 844 | 6816957 | 99 | ||
| BC059048.1 | - | 844 | 6816957 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem163
Genomic Map