Assay Details
| Gene Symbol: Ccnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028399.1 | NP_082675.1 | 2 - 3 | 298 | 96 | |
| GenBank mRNA | AK013634.1 | - | 2 - 3 | 245 | 96 | |
| AK036772.1 | - | 246 | 96 | |||
| AK049863.1 | - | 2 - 3 | 243 | 96 | ||
| AK152399.1 | - | 2 - 3 | 246 | 96 | ||
| AK154439.1 | - | 2 - 3 | 256 | 96 | ||
| AK156231.1 | - | 267 | 96 | |||
| AK156621.1 | - | 2 - 3 | 254 | 96 | ||
| BC054122.1 | - | 2 - 3 | 298 | 6509024 | 96 | |
| JX965957.1 | - | 2 - 3 | 234 | 96 | ||
| JX965958.1 | - | 2 - 3 | 234 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccnt2
Genomic Map