Assay Details
| Gene Symbol: Ccnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028399.1 | NP_082675.1 | 6 - 7 | 605 | 80 | |
| GenBank mRNA | AK036772.1 | - | 553 | 80 | ||
| AK049863.1 | - | 6 - 7 | 550 | 80 | ||
| AK152399.1 | - | 6 - 7 | 553 | 80 | ||
| AK154439.1 | - | 6 - 7 | 563 | 80 | ||
| AK156231.1 | - | 574 | 80 | |||
| BC054122.1 | - | 6 - 7 | 605 | 6509024 | 80 | |
| JX965957.1 | - | 6 - 7 | 541 | 80 | ||
| JX965958.1 | - | 6 - 7 | 541 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccnt2
Genomic Map