Assay Details
| Gene Symbol: Acmsd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033041.2 | NP_001028213.1 | 4 - 5 | 326 | 71 | |
| GenBank mRNA | AB071419.1 | - | 4 - 5 | 253 | 71 | |
| AK143962.1 | - | 4 - 5 | 328 | 71 | ||
| BC139281.1 | - | 4 - 5 | 277 | 8862303 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acmsd
Genomic Map