Assay Details
| Gene Symbol: Gm19990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045048.1 | 1 - 2 | 92 | 80 | ||
| GenBank mRNA | AK053248.1 | - | 1 - 2 | 93 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | breast cancer metastasis-suppressor 1-like | Brms1l |
0710008O11Rik, AI159718, BRMS1, D12Ertd407e |
-383.5kb | Sense | Chr. 12 - 55836243 - 55869736 on NCBI Build GRCm38 |
| Downstream | MAP3K12 binding inhibitory protein 1 | Mbip | 4933408E06Rik | 66.8kb | Anti-sense | Chr. 12 - 56328306 - 56345894 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm19990
Genomic Map