Assay Details
| Gene Symbol: Mgat5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145128.3 | NP_660110.2 | 9 - 10 | 1607 | 77 | |
| GenBank mRNA | AF474154.1 | - | 7 - 8 | 987 | 77 | |
| AF474155.1 | - | 7 - 8 | 1050 | 77 | ||
| AK030402.1 | - | 9 - 10 | 1575 | 77 | ||
| AK036567.1 | - | 9 - 10 | 1562 | 77 | ||
| BC125533.1 | - | 7 - 8 | 1015 | 40130048 | 77 | |
| BC125535.1 | - | 7 - 8 | 1015 | 40130050 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mgat5
Genomic Map