Assay Details
| Gene Symbol: Tnfsf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183391.3 | NP_899247.3 | 1 - 2 | 194 | 72 | |
| GenBank mRNA | AJ577579.1 | - | 1 - 2 | 196 | 72 | |
| AJ577580.1 | - | 196 | 72 | |||
| AY234223.1 | - | 1 - 2 | 152 | 72 | ||
| AY320040.1 | - | 1 - 2 | 144 | 72 | ||
| AY359852.1 | - | 1 - 2 | 138 | 72 | ||
| BC137814.1 | - | 1 - 2 | 194 | 8860833 | 72 | |
| BC137815.1 | - | 1 - 2 | 194 | 8860834 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tnfsf18
Genomic Map