Assay Details
| Gene Symbol: Ahr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF405560.1 | - | 10 - 11 | 2413 | 72 | |
| AF405561.1 | - | 10 - 11 | 2413 | 72 | ||
| AF405562.1 | - | 10 - 11 | 2413 | 72 | ||
| AF405565.1 | - | 10 - 11 | 2413 | 72 | ||
| AF405566.1 | - | 10 - 11 | 2413 | 72 | ||
| AF405567.1 | - | 10 - 11 | 2413 | 72 | ||
| AF405568.1 | - | 10 - 11 | 72 | |||
| AF405570.1 | - | 10 - 11 | 2413 | 72 | ||
| AY662085.1 | - | 1 - 2 | 1156 | 72 | ||
| AY662086.1 | - | 1 - 2 | 1161 | 72 | ||
| AY662087.1 | - | 1 - 2 | 1156 | 72 | ||
| AY662088.1 | - | 1 - 2 | 1156 | 72 | ||
| AY662089.1 | - | 1 - 2 | 1156 | 72 | ||
| AY662090.1 | - | 1 - 2 | 1156 | 72 | ||
| BC040248.1 | - | 1 - 2 | 316 | 72 | ||
| D38416.1 | - | 10 - 11 | 2428 | 72 | ||
More Information
Set Membership: |
|

Ahr
Genomic Map