Assay Details
| Gene Symbol: Serpinb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198028.3 | NP_932145.3 | 3 - 4 | 335 | 118 | |
| GenBank mRNA | AK036537.1 | - | 3 - 4 | 313 | 118 | |
| BC029736.1 | - | 3 - 4 | 276 | 3676050 | 118 | |
| BC069938.1 | - | 3 - 4 | 295 | 5035202 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Serpinb10
Genomic Map