Assay Details
| Gene Symbol: Agr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011783.2 | NP_035913.1 | 3 - 4 | 259 | 85 | |
| GenBank mRNA | AB016592.1 | - | 3 - 4 | 258 | 85 | |
| AF044262.1 | - | 3 - 4 | 259 | 85 | ||
| AK007677.1 | - | 3 - 4 | 278 | 85 | ||
| BC013334.1 | - | 3 - 4 | 277 | 4218549 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Agr2
Genomic Map