Assay Details
| Gene Symbol: Agr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207531.3 | NP_997414.2 | 7 - 8 | 579 | 151 | |
| GenBank mRNA | AK053177.1 | - | 7 - 8 | 508 | 151 | |
| AW490945.1 | - | 1 - 2 | 151 | |||
| BC023499.1 | - | 7 - 8 | 580 | 5008294 | 151 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P433789 | BC023499.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P433789 | NM_207531.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P433789 | AK053177.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P433789 | AW490945.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P433789 | BC023499.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P433789 | NM_207531.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P433789 | AK053177.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P433789 | AW490945.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 180598 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 809883 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 163524 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 125858 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 342777 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 190517 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 276047 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 492879 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 601935 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 462168 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 465501 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 78262 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 38250 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 146524 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 172616 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155975 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177930 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P433789 | BC023499.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P433789 | NM_207531.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P433789 | AK053177.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P433789 | AW490945.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P433789 | BC023499.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P433789 | NM_207531.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P433789 | AK053177.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P433789 | AW490945.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28355521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28355556 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28355532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28355548 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28355543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28355527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28355530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28355536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28355549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28355542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28355526 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28355545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28355552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28355537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28355535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28355554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28355524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28355539 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28355546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28355544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28355533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28355529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28355523 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155975 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07587840 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177930 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07587849 | - | - |
| Whole Mouse Genome G4122A | A_51_P433789 | BC023499.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433789 | NM_207531.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433789 | AK053177.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433789 | AW490945.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433789 | BC023499.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433789 | NM_207531.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433789 | AK053177.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433789 | AW490945.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433796 | BC023499.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433796 | NM_207531.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433796 | AK053177.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433796 | AW490945.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127318 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155975 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177930 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09943172 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09943167 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10031381 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09943171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28355568 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28355502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28355505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28355526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28355509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10399377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28355515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28355521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28355513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28355535 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10194817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28355495 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28355536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28355533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28355499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28355512 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28355517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28355523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28355527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28355519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28355504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28355498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28355516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28355507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28355530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10235372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10399375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28355532 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28355496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09859271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28355511 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10395357 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395360 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395362 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395363 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395364 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395359 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395361 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395358 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425221_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10395356 | - | - |
| Mouse Genome 430 2.0 Array | 1425221_at | - | - |
| Mouse Genome 430 2.0 Array | 1425221_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 403205_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 403205_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200002367.mm.1 | NM_207531.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200002367.mm.1 | NM_207531.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200002367.mm.1 | TC1200000314.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425221_at | - | - |
| Mouse Expression Set 430 | 1425221_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425221_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425221_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425221_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425221_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 298656 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2340008 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002340008 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1215701 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1215701 | - | - |
| mouse-6 v1.1 (Illumina) | scl0403205.5_11-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2340008 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0403205.5_11-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2340008 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2902353 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2902353 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1215701 | - | - |

Agr3
Genomic Map