Assay Details
| Gene Symbol: Vangl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033509.3 | NP_277044.1 | 1 - 2 | 112 | ||
| GenBank mRNA | AK033128.1 | - | 1 - 2 | 112 | ||
| AK046036.1 | - | 1 - 2 | 112 | |||
| AK129311.1 | - | 272 | 112 | |||
| AY035370.3 | - | 1 - 2 | 112 | |||
| BC052195.1 | - | 1 - 2 | 272 | 6509008 | 112 | |
| BC075636.1 | - | 112 | ||||
More Information
Additional Information:
This assay is in the 5'UTR region of the AY035370.3, BC052195.1, NM_033509.3 transcripts.
Set Membership: |
|

Vangl2
Genomic Map