Assay Details
| Gene Symbol: Ankmy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146033.3 | NP_666145.3 | 7 - 8 | 1116 | 72 | |
| GenBank mRNA | AK037644.1 | - | 5 - 6 | 3875 | 72 | |
| AK077556.1 | - | 7 - 8 | 1087 | 72 | ||
| AK164434.1 | - | 7 - 8 | 1018 | 72 | ||
| BC024959.1 | - | 7 - 8 | 1073 | 4946879 | 72 | |
More Information
Set Membership: |
|

Ankmy2
Genomic Map