Assay Details
| Gene Symbol: 1700112D23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007174.1 | - | 171 | 63 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 2 (facilitated glucose transporter), member 2 |
Slc2a2 |
AI266973, Glut-2, Glut2 |
-13.8kb | Anti-sense | Chr. 3 - 28697903 - 28728361 on NCBI Build GRCm38 |
| Downstream | eukaryotic translation initiation factor 5A2 | Eif5a2 |
9630038B20, eIF5AII |
611.0 | Anti-sense | Chr. 3 - 28781311 - 28798846 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

1700112D23Rik
Genomic Map