Assay Details
Gene Symbol: Atxn7l1os2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK016151.1 | - | 166 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ataxin 7-like 1 | Atxn7l1 |
2810423G08Rik, Atxn7l4 |
Anti-sense | Chr. 12 - 33147639 - 33376408 on NCBI Build GRCm38 | |
Downstream | EF-hand calcium binding domain 10 | Efcab10 | 4930504H06Rik | 10.9kb | Anti-sense | Chr. 12 - 33394854 - 33401269 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() |
