Assay Details
| Gene Symbol: 2610100L16Rik | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033490.1 | 1 - 2 | 263 | 83 | |||
| GenBank mRNA | AK011787.1 | - | LIT3367 2610100L16 |
1 - 2 | 238 | 83 | |
| AK163433.1 | - | 1 - 2 | 136 | 83 | |||
| BE647934.1 | - | 1 - 2 | 263 | 83 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 30340 |
Gm30340 | -4.0kb | Anti-sense | Chr. 3 - 17287413 - 17786005 on NCBI Build GRCm38 | |
| Downstream | microRNA 124a-2 | Mir124a-2 |
Mirn124a-2, mir-124-2, mir-124a-2 |
901.0 | Sense | Chr. 3 - 17795662 - 17795770 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

2610100L16Rik
Genomic Map