Assay Details
| Gene Symbol: Dars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177445.5 | NP_803228.2 | 11 - 12 | 1305 | 70 | |
| GenBank mRNA | AK077533.1 | - | 11 - 12 | 1228 | 70 | |
| AK144470.1 | - | 11 - 12 | 1251 | 70 | ||
| AK146350.1 | - | 11 - 12 | 1249 | 70 | ||
| AK151027.1 | - | 11 - 12 | 1249 | 70 | ||
| AK151897.1 | - | 11 - 12 | 1216 | 70 | ||
| AK152304.1 | - | 11 - 12 | 1216 | 70 | ||
| AK152562.1 | - | 11 - 12 | 1216 | 70 | ||
| AK153256.1 | - | 11 - 12 | 1216 | 70 | ||
| AK153299.1 | - | 11 - 12 | 1216 | 70 | ||
| AK166009.1 | - | 11 - 12 | 1216 | 70 | ||
| AK169247.1 | - | 11 - 12 | 1216 | 70 | ||
| AK169716.1 | - | 11 - 12 | 1193 | 70 | ||
| BC008638.1 | - | 11 - 12 | 1203 | 3586278 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dars
Genomic Map