Assay Details
| Gene Symbol: Mcm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008567.1 | NP_032593.1 | 12 - 13 | 1840 | 107 | |
| GenBank mRNA | AK078126.1 | - | 6 - 7 | 834 | 107 | |
| AK088493.1 | - | 12 - 13 | 1839 | 107 | ||
| AK145520.1 | - | 12 - 13 | 1831 | 107 | ||
| AK146770.1 | - | 12 - 13 | 1833 | 107 | ||
| AK146896.1 | - | 12 - 13 | 1715 | 107 | ||
| AK150726.1 | - | 12 - 13 | 1788 | 107 | ||
| AK198870.1 | - | 2 - 3 | 165 | 107 | ||
| AK217569.1 | - | 2 - 3 | 165 | 107 | ||
| BC050886.1 | - | 12 - 13 | 1848 | 5703922 | 107 | |
| BC051658.1 | - | 12 - 13 | 1815 | 107 | ||
| BC057584.1 | - | 12 - 13 | 1818 | 6401827 | 107 | |
| D86726.1 | - | 12 - 13 | 1840 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mcm6
Genomic Map